Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLX2
Entrez Gene ID
709649
Gene Name
T cell leukemia homeobox 2
Gene Aliases
-
Chromosome Location
Chr.13: 34299230 - 34301939 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001110250.4 XP_001110250.1 - 757 128

View Full Product Details