Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REEP6
Entrez Gene ID
707662
Gene Name
receptor accessory protein 6
Gene Aliases
-
Chromosome Location
Chr.19: 1285419 - 1292759 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194035.2 NP_001180964.1 - 450 78
GenBank mRNA JV048710.1 - - 309 78
JV636458.1 - - 287 78

View Full Product Details