Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STOM
Entrez Gene ID
699584
Gene Name
stomatin
Gene Aliases
-
Chromosome Location
Chr.15: 21342828 - 21375662 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266229.1 NP_001253158.1 - 566 98
XM_028834248.1 XP_028690081.1 - 524 98
GenBank mRNA JU333342.1 - - 594 98
JU474642.1 - - 594 98
JV045309.1 - - 566 98
JV565288.1 - - 598 98

View Full Product Details