Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTHLH
Entrez Gene ID
710295
Gene Name
parathyroid hormone like hormone
Gene Aliases
EGK_03491
Chromosome Location
Chr.11: 29283410 - 29296069 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261680.1 NP_001248609.1 - 103 104
XM_015151150.2 XP_015006636.1 - 495 104
XM_015151151.2 XP_015006637.1 - 836 104
GenBank mRNA JU322180.1 - - 103 104

View Full Product Details