Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF12
Entrez Gene ID
694575
Gene Name
Kruppel like factor 12
Gene Aliases
EGK_09372
Chromosome Location
Chr.17: 55480829 - 55925505 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258129.1 NP_001245058.1 - 777 106
XM_015121180.2 XP_014976666.1 - 1144 106
GenBank mRNA JU327687.1 - - 628 106
JU471196.1 - - 777 106

View Full Product Details