Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC715304
Entrez Gene ID
715304
Gene Name
lymphocyte antigen 6 complex locus protein G6d
Gene Aliases
g6g-ly6g6d, ly6g6d, ly6g6f
Chromosome Location
Chr.4: 137785100 - 137798352 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001105454.4 XP_001105454.2 - 615 89
XM_015135991.2 XP_014991477.1 - 740 89
XM_028847719.1 XP_028703552.1 - 722 89
GenBank mRNA KU253645.1 - - 418 89
KU253646.1 - - 919 89
KU253647.1 - - 418 89
KU253648.1 - - 418 89
KU253649.1 - - 418 89

View Full Product Details