Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ODF1
Entrez Gene ID
707269
Gene Name
outer dense fiber of sperm tails 1
Gene Aliases
EGK_19181
Chromosome Location
Chr.8: 102819251 - 102828942 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100871.4 XP_001100871.1 - 479 85

View Full Product Details