Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL15
Entrez Gene ID
698391
Gene Name
kelch like family member 15
Gene Aliases
EGK_20331
Chromosome Location
Chr.X: 23797166 - 23844513 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU471989.1 - - 96 129
JV048309.1 - - 148 129

View Full Product Details