Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMAT1
Entrez Gene ID
706436
Gene Name
zinc finger matrin-type 1
Gene Aliases
-
Chromosome Location
Chr.X: 98643598 - 98691874 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015127836.2 XP_014983322.1 - 961 133
XM_015127837.2 XP_014983323.1 - 958 133
XM_015127838.2 XP_014983324.1 - 7672 133
XM_015127839.2 XP_014983325.1 - 2134 133
XM_028842437.1 XP_028698270.1 - 892 133
GenBank mRNA JU349547.1 - - 96 133
JV725169.1 - - 95 133

View Full Product Details