Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPAM1
Entrez Gene ID
696249
Gene Name
sperm adhesion molecule 1
Gene Aliases
-
Chromosome Location
Chr.3: 150338424 - 150372830 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086882.4 XP_001086882.2 - 227 167
XM_015134834.2 XP_014990320.1 - 300 167

View Full Product Details