Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMCC1
Entrez Gene ID
702576
Gene Name
transmembrane and coiled-coil domain family 1
Gene Aliases
EGK_03997
Chromosome Location
Chr.2: 146578982 - 146832518 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261150.1 NP_001248079.1 - 1804 114
XM_015130128.2 XP_014985614.1 - 2163 114
XM_015130129.2 XP_014985615.1 - 2169 114
XM_015130130.2 XP_014985616.1 - 2110 114
XM_015130131.2 XP_014985617.1 - 1732 114
XM_015130132.2 XP_014985618.1 - 1655 114
XM_015130133.2 XP_014985619.1 - 2273 114
XM_028843187.1 XP_028699020.1 - 2315 114
XM_028843188.1 XP_028699021.1 - 1909 114
XM_028843189.1 XP_028699022.1 - 1421 114
XM_028843191.1 XP_028699024.1 - 1478 114
XM_028843192.1 XP_028699025.1 - 2288 114
XM_028843193.1 XP_028699026.1 - 1843 114
GenBank mRNA JU320319.1 - - 2068 114
JU320320.1 - - 2068 114
JU473049.1 - - 1968 114
JU473050.1 - - 1968 114
JV044513.1 - - 1804 114
JV044514.1 - - 1804 114
JV645848.1 - - 2071 114

View Full Product Details