Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASIC3
Entrez Gene ID
714396
Gene Name
acid sensing ion channel subunit 3
Gene Aliases
ACCN3
Chromosome Location
Chr.3: 177049289 - 177054756 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001102336.4 XP_001102336.1 - 1800 116
GenBank mRNA JU330873.1 - - 489 116

View Full Product Details