Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC4A
Entrez Gene ID
716122
Gene Name
C-type lectin domain family 4 member A
Gene Aliases
EGK_03325
Chromosome Location
Chr.11: 11382672 - 11403401 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001113483.4 XP_001113483.1 - 1634 96
XM_028829255.1 XP_028685088.1 - 1466 96

View Full Product Details