Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE1
Entrez Gene ID
693690
Gene Name
jade family PHD finger 1
Gene Aliases
EGK_16088, PHF17
Chromosome Location
Chr.5: 126580612 - 126649058 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266133.1 NP_001253062.1 - 462 119
XM_015138933.2 XP_014994419.1 - 343 119
XM_015138934.2 XP_014994420.1 - 714 119
XM_015138935.2 XP_014994421.1 - 451 119
XM_015138936.2 XP_014994422.1 - 533 119
XM_028847943.1 XP_028703776.1 - 425 119
XM_028847944.1 XP_028703777.1 - 458 119
GenBank mRNA JU336457.1 - - 425 119
JU477273.1 - - 428 119
JV048293.1 - - 462 119
JV661899.1 - - 556 119

View Full Product Details