Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMC10
Entrez Gene ID
719285
Gene Name
ER membrane protein complex subunit 10
Gene Aliases
C19H19orf63, C19orf63
Chromosome Location
Chr.19: 50383940 - 50391420 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194699.2 NP_001181628.1 - 441 124
XM_015124415.2 XP_014979901.1 - 506 124
XM_015124417.2 XP_014979903.1 - 993 124
GenBank mRNA JU322500.1 - - 487 124
JU484832.1 - - 124
JV045277.1 - - 481 124
JV579089.1 - - 490 124

View Full Product Details