Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXLNA
Entrez Gene ID
707704
Gene Name
taxilin alpha
Gene Aliases
EGK_00498
Chromosome Location
Chr.1: 192469965 - 192489489 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015133929.2 XP_014989415.1 - 1242 85
XM_028836515.1 XP_028692348.1 - 1326 85
XM_028836522.1 XP_028692355.1 - 1528 85
GenBank mRNA JU333299.1 - - 1157 85
JU474612.1 - - 1104 85

View Full Product Details