Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19H19orf47
Entrez Gene ID
701191
Gene Name
chromosome 19 C19orf47 homolog
Gene Aliases
-
Chromosome Location
Chr.19: 39526156 - 39555142 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001095873.4 XP_001095873.1 - 206 108
XM_028840197.1 XP_028696030.1 - 200 108
GenBank mRNA JU332154.1 - - 170 108

View Full Product Details