Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LDOC1
Entrez Gene ID
696902
Gene Name
LDOC1 regulator of NFKB signaling
Gene Aliases
EGK_21009
Chromosome Location
Chr.X: 137457070 - 137458419 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260647.2 NP_001247576.1 - 54 137
GenBank mRNA JU321914.1 - - 109 137
JV044712.1 - - 55 137

View Full Product Details