Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLUL
Entrez Gene ID
715952
Gene Name
glutamate-ammonia ligase
Gene Aliases
-
Chromosome Location
Chr.1: 39623412 - 39629245 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266013.2 NP_001252942.1 - 373 132
GenBank mRNA JU471458.1 - - 670 132
JU475223.1 - - 698 132
JV043358.1 - - 472 132
JV465152.1 - - 511 132
JV635548.1 - - 373 132

View Full Product Details