Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD14
Entrez Gene ID
697482
Gene Name
CD14 molecule
Gene Aliases
-
Chromosome Location
Chr.6: 138012823 - 138014253 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130433.2 NP_001123905.1 - 939 63
GenBank mRNA AB446511.1 - - 848 63
FS723629.1 - - 488 63
JU325836.1 - - 504 63
JU473288.1 - - 853 63

View Full Product Details