Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIR30E
Entrez Gene ID
100315186
Gene Name
microRNA mir-30e
Gene Aliases
mir-30e, mml-mir-30e
Chromosome Location
Chr.1: 183824813 - 183824904 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_032384.1 - 46 83

View Full Product Details