The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053507.1 | NP_445959.1 | 2 - 3 | 181 | 60 |
XM_008767598.1 | XP_008765820.1 | 2 - 3 | 293 | 60 | |
GenBank mRNA | AY017337.1 | - | 2 - 3 | 181 | 60 |