Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gng7
Entrez Gene ID
58979
Gene Name
guanine nucleotide binding protein (G protein), gamma 7
Gene Aliases
-
Chromosome Location
Chr.7: 11582984 - 11628929 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024138.1 NP_077052.1 2 - 3 325 75
GenBank mRNA DQ120497.1 - 1 - 2 83 75
DQ120498.1 - 1 - 2 83 75
FQ211653.1 - - 305 75
L23219.1 - 2 - 3 325 75

View Full Product Details