The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053550.1 | NP_446002.1 | 7 - 8 | 919 | 96 |
XM_008768131.1 | XP_008766353.1 | 7 - 8 | 1198 | 96 | |
GenBank mRNA | AF033355.1 | - | 7 - 8 | 919 | 96 |