The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053967.2 | NP_446419.1 | 3 - 4 | 1370 | 132 |
GenBank mRNA | BC081748.1 | - | 3 - 4 | 1311 | 132 |
X89999.1 | - | 3 - 4 | 1374 | 132 |