The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057101.2 | NP_476442.2 | 7 - 8 | 937 | 74 |
XM_006255907.2 | XP_006255969.1 | 7 - 8 | 1011 | 74 | |
GenBank mRNA | U56853.1 | - | 7 - 8 | 938 | 74 |
U56854.1 | - | 6 - 7 | 999 | 74 |