The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153734.1 | NP_714956.1 | 2 - 3 | 352 | 112 |
GenBank mRNA | AF440736.1 | - | 2 - 3 | 352 | 112 |