Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vom2r-ps45
Entrez Gene ID
286982
Gene Name
vomeronasal 2 receptor, pseudogene 45
Gene Aliases
-
Chromosome Location
Chr.1: 69496423 - 69522162 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF016183.1 - - 510 106

View Full Product Details