Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plat
Entrez Gene ID
25692
Gene Name
plasminogen activator, tissue
Gene Aliases
PATISS, tPA
Chromosome Location
Chr.16: 74098263 - 74122897 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013151.2 NP_037283.2 8 - 9 906 63
XM_006253347.2 XP_006253409.1 8 - 9 978 63
XM_008771348.1 XP_008769570.1 9 - 10 1036 63
GenBank mRNA BC061565.1 - 8 - 9 906 63
M23697.1 - 7 - 8 830 63

Gene Symbol
LOC100910418
Entrez Gene ID
100910418
Gene Name
tissue-type plasminogen activator-like
Gene Aliases
-
Chromosome Location
Chr.16: 73745773 - 73763212 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751623.3 XP_003751671.2 8 - 9 910 63

View Full Product Details