The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013200.1 | NP_037332.1 | 11 - 12 | 1387 | 83 |
XM_006242180.2 | XP_006242242.1 | 11 - 12 | 1843 | 83 | |
GenBank mRNA | BC085761.1 | - | 11 - 12 | 1399 | 83 |
D43623.1 | - | 10 - 11 | 1374 | 83 |