The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080580.2 | NP_542147.1 | 4 - 5 | 580 | 81 |
XM_006242593.1 | XP_006242655.1 | 4 - 5 | 576 | 81 | |
XM_006242594.2 | XP_006242656.1 | 4 - 5 | 696 | 81 | |
GenBank mRNA | BC081741.1 | - | 4 - 5 | 580 | 81 |
M83681.1 | - | 3 - 4 | 408 | 81 | |
U90206.1 | - | 3 - 4 | 471 | 81 |