Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ikzf2
Entrez Gene ID
301476
Gene Name
IKAROS family zinc finger 2
Gene Aliases
Zfpn1a2
Chromosome Location
Chr.9: 76621230 - 76770485 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106916.1 NP_001100386.1 4 - 5 565 78
XM_006245119.2 XP_006245181.1 4 - 5 711 78
XM_006245120.2 XP_006245182.1 4 - 5 459 78
XM_006245121.2 XP_006245183.1 4 - 5 606 78
XM_008767217.1 XP_008765439.1 4 - 5 560 78
XM_008767218.1 XP_008765440.1 4 - 5 538 78

View Full Product Details