Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfic
Entrez Gene ID
29228
Gene Name
nuclear factor I/C
Gene Aliases
-
Chromosome Location
Chr.7: 11151952 - 11179544 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031567.1 NP_113755.1 9 - 10 1352 95
GenBank mRNA AB012233.2 - 9 - 10 1352 95

View Full Product Details