Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sirt6
Entrez Gene ID
299638
Gene Name
sirtuin 6
Gene Aliases
-
Chromosome Location
Chr.7: 10937622 - 10943048 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031649.1 NP_001026819.1 4 - 5 464 70
XM_006240896.1 XP_006240958.1 5 - 6 621 70
XM_006240897.2 XP_006240959.1 4 - 5 1090 70
XM_008765091.1 XP_008763313.1 4 - 5 1115 70
XM_008765092.1 XP_008763314.1 5 - 6 860 70
GenBank mRNA BC098923.1 - 4 - 5 464 70

View Full Product Details