Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ribc1
Entrez Gene ID
317431
Gene Name
RIB43A domain with coiled-coils 1
Gene Aliases
-
Chromosome Location
Chr.X: 21699361 - 21711345 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007715.1 NP_001007716.1 3 - 4 284 71
XM_006256799.2 XP_006256861.1 2 - 3 471 71
XM_006256800.2 XP_006256862.1 3 - 4 424 71
XM_006256801.2 XP_006256863.1 3 - 4 435 71
XM_006256802.2 XP_006256864.1 3 - 4 339 71
XM_006256803.2 XP_006256865.1 3 - 4 342 71
XM_006256804.2 XP_006256866.1 3 - 4 324 71
XM_006256805.2 XP_006256867.1 3 - 4 320 71
GenBank mRNA BC078998.1 - 3 - 4 284 71

View Full Product Details