The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105982.1 | NP_001099452.1 | 8 - 9 | 1615 | 115 |
XM_006250373.2 | XP_006250435.1 | 8 - 9 | 1775 | 115 | |
XM_006250375.2 | XP_006250437.1 | 8 - 9 | 1654 | 115 | |
GenBank mRNA | FQ212123.1 | - | 8 - 9 | 1599 | 115 |