Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem27
Entrez Gene ID
57395
Gene Name
transmembrane protein 27
Gene Aliases
NX-17, Nx17, collectrin
Chromosome Location
Chr.X: 32118082 - 32153687 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC078838.1 - 1 - 2 118 123

View Full Product Details