Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mutyh
Entrez Gene ID
170841
Gene Name
mutY homolog
Gene Aliases
Myh
Chromosome Location
Chr.5: 135663332 - 135675348 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133316.1 NP_579850.1 10 - 11 123
XM_006238626.2 XP_006238688.1 10 - 11 932 123
XM_006238627.2 XP_006238689.1 9 - 10 829 123
XM_008763987.1 XP_008762209.1 8 - 9 890 123
GenBank mRNA AF478683.1 - 10 - 11 123

View Full Product Details