Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtfmt
Entrez Gene ID
315763
Gene Name
mitochondrial methionyl-tRNA formyltransferase
Gene Aliases
RGD1309462
Chromosome Location
Chr.8: 70952173 - 70986393 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009697.1 NP_001009697.1 7 - 8 898 110
GenBank mRNA BC088470.1 - 7 - 8 898 110
FQ226710.1 - 3 - 4 225 110

View Full Product Details