Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cope
Entrez Gene ID
290659
Gene Name
coatomer protein complex, subunit epsilon
Gene Aliases
-
Chromosome Location
Chr.16: 20863141 - 20873521 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106076.1 NP_001099546.1 1 - 2 333 77
XM_006252876.2 XP_006252938.1 1 - 2 347 77
GenBank mRNA BC161928.1 - 1 - 2 347 77
FQ220757.1 - 1 - 2 157 77
FQ232202.1 - 1 - 2 156 77
FQ233163.1 - 1 - 2 156 77

View Full Product Details