The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004081.1 | NP_001004081.1 | 6 - 7 | 859 | 57 |
GenBank mRNA | BC079111.1 | - | 6 - 7 | 859 | 57 |