The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012124.1 | NP_001012124.1 | 1 - 1 | 1305 | 61 |
GenBank mRNA | BC087592.1 | - | 1 - 1 | 1305 | 61 |