Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pigt
Entrez Gene ID
296360
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class T
Gene Aliases
-
Chromosome Location
Chr.3: 160944293 - 160954738 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106540.2 NP_001100010.2 1 - 2 222 67
XM_006235532.2 XP_006235594.1 1 - 2 1508 67

View Full Product Details