Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agpat3
Entrez Gene ID
294324
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases
-
Chromosome Location
Chr.20: 11060647 - 11144806 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106378.1 NP_001099848.1 3 - 4 740 81
XM_006256243.2 XP_006256305.1 4 - 5 535 81
XM_006256244.2 XP_006256306.1 4 - 5 563 81
GenBank mRNA BC158873.1 - 2 - 3 290 81

View Full Product Details