Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mapk13
Entrez Gene ID
29513
Gene Name
mitogen activated protein kinase 13
Gene Aliases
Prkm13
Chromosome Location
Chr.20: 6018374 - 6027467 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019231.2 NP_062104.2 9 - 10 822 93
GenBank mRNA AF092534.1 - 9 - 10 764 93
BC107849.1 - 9 - 10 822 93

View Full Product Details