Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usp1
Entrez Gene ID
313387
Gene Name
ubiquitin specific peptidase 1
Gene Aliases
-
Chromosome Location
Chr.5: 117583502 - 117594872 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015015.1 NP_001015015.1 6 - 7 1702 137
GenBank mRNA BC092574.1 - 6 - 7 1702 137

View Full Product Details