The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139483.1 | NP_001132955.1 | 7 - 8 | 993 | 59 |
XM_006256452.2 | XP_006256514.1 | 8 - 9 | 1110 | 59 | |
XM_006256453.2 | XP_006256515.1 | 8 - 9 | 1107 | 59 | |
XM_006256454.2 | XP_006256516.1 | 8 - 9 | 1053 | 59 | |
XM_006256456.2 | XP_006256518.1 | 8 - 9 | 1080 | 59 | |
XM_008772888.1 | XP_008771110.1 | 8 - 9 | 1110 | 59 | |
XM_008772889.1 | XP_008771111.1 | 8 - 9 | 1068 | 59 | |
XM_008772890.1 | XP_008771112.1 | 8 - 9 | 1055 | 59 | |
GenBank mRNA | CF114906.1 | - | 4 - 5 | 567 | 59 |