Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ifnk
Entrez Gene ID
313152
Gene Name
interferon kappa
Gene Aliases
-
Chromosome Location
Chr.5: 50631217 - 50634142 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107925.1 NP_001101395.1 1 - 2 575 125

View Full Product Details