Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmd5
Entrez Gene ID
296651
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 5
Gene Aliases
-
Chromosome Location
Chr.3: 13916199 - 13937038 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106569.2 NP_001100039.2 2 - 3 337 72
XM_006233969.2 XP_006234031.1 2 - 3 866 72
XM_006233970.2 XP_006234032.1 3 - 4 956 72
GenBank mRNA BC166850.1 - 2 - 3 319 72

View Full Product Details