Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lcn3
Entrez Gene ID
502611
Gene Name
lipocalin 3
Gene Aliases
RGD1562745
Chromosome Location
Chr.3: 2945312 - 2947318 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008761616.1 XP_008759838.1 4 - 5 425 67

View Full Product Details